Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

16817

Sigma-Aldrich

S-Ipsdienol

≥97.0% (HPLC)

Synonym(s):

(S)-(+)-2-Methyl-6-methylene-2,7-octadien-4-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
MDL number:
UNSPSC Code:
12352200

assay

≥97.0% (HPLC)

optical activity

[α]/D +16±3°, c = 1 in methanol

storage temp.

2-8°C

SMILES string

C\C(C)=C\[C@@H](O)CC(=C)C=C

Application

S-Ipsdienol has been used to study the genetic control of the enantiomeric composition of ipsdienol in the pine engraver, Ips pini. S-Ipsdienol has also been used to determine that myrcene hydroxylases do not determine enantiomeric composition of pheromonal ipsdienol in Ips spp.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Environment

signalword

Warning

hcodes

pcodes

Hazard Classifications

Aquatic Acute 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

188.6 °F

flash_point_c

87 °C

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Seybold et al.
Journal of chemical ecology, 21(7), 995-1016 (1995-07-01)
Thirty-five populations ofIps pini (Say) and one population each ofIps avulsus (Eichhoff) andIps bonanseai (Hopkins) were analyzed for the enantiomeric composition of ipsdienol (2-methyl-6-methylene-2,7-octadien-4-ol). Populations ofI. pini occur as at least two distinct regional pheromone variants: New York type [32%-(-)
Pheromonal chirality and integrity of aggregation response in southern species of the bark beetle Ips sp.
Vite, J.P., et al.
Nature, 272, 817-818 (1978)
Synergistic effect of a pheromone and a kairomone on host selection and colonisation by Ips avulsus.
R Hedden et al.
Nature, 261(5562), 696-697 (1976-06-24)
Pamela Sandstrom et al.
Journal of chemical ecology, 34(12), 1584-1592 (2008-11-27)
Myrcene (7-methyl-3-methylene-1,6-octadiene) hydroxylation is likely one of the final reactions involved in the production of the Ips spp. (Coleoptera: Scolytidae) aggregation pheromone components, ipsdienol (2-methyl-6-methylene-2,7-octadien-4-ol) and ipsenol (2-methyl-6-methylene-7-octen-4-ol). To gain insight into the evolution of pheromone production, we isolated a
Michael J Domingue et al.
Journal of chemical ecology, 32(5), 1005-1026 (2006-06-02)
The genetic nature of pheromone variation within species has rarely been studied, and never for male-produced long-range pheromones. Males from western North American populations of Ips pini produce predominantly (R)-(-)-ipsdienol, whereas those from eastern North American populations produce higher proportions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service