Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

71872

Sigma-Aldrich

Sodium (para)periodate

purum p.a., ≥99.0% (as H2INa3O6, RT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na3H2IO6
CAS Number:
Molecular Weight:
293.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
≥99.0% (as H2INa3O6, RT)
form:
powder

grade

purum p.a.

assay

≥99.0% (as H2INa3O6, RT)

form

powder

reaction suitability

reagent type: oxidant

pH

3.5-5.5 (25 °C, 106.9 g/L)

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤500 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Na+].[Na+].[Na+].OI(O)([O-])([O-])([O-])=O

InChI

1S/H5IO6.3Na/c2-1(3,4,5,6)7;;;/h(H5,2,3,4,5,6,7);;;/q;3*+1/p-3

InChI key

DSYMXPNUZLZKOA-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Sodium (para)periodate is a hypervalent iodine compound.

pictograms

Flame over circleExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service