Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W470620

Sigma-Aldrich

9-decen-2-one

≥95%

Synonym(s):

dec-9-en-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
FEMA Number:
4706
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
07.262
NACRES:
NA.21
organoleptic:
apple; fatty; green; fruity; pear; pineapple
biological source:
synthetic

biological source

synthetic

Quality Level

assay

≥95%

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

organoleptic

apple; fatty; green; fruity; pear; pineapple

SMILES string

CC(CCCCCCC=C)=O

InChI

1S/C10H18O/c1-3-4-5-6-7-8-9-10(2)11/h3H,1,4-9H2,2H3

InChI key

DZECLZDTSQJBSU-UHFFFAOYSA-N

General description

9-Decen-2-one is an aliphatic ketone. It has been identified as one of the volatile components present in the normal cork samples and in the cork samples infected with Armillaria mellea and by molds.[1] It is also present in the fresh pineapple fruits (Ananas comosus).[2]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

141.8 °F

flash_point_c

61 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GC-MS study of volatiles of normal and microbiologically attacked cork from Quercus suber L.
Rocha S, et al.
Journal of Agricultural and Food Chemistry, 44(3), 865-871 (1996)
Identification of 9-decen-2-one in pineapple.
Zucca J and Schippa C.
Perfumer & Flavorist, 34(7), 42-48 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service