Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W470301

Sigma-Aldrich

Cinnamyl benzoate

≥98%, FG

Synonym(s):

3-Phenyl-2-propenyl benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O2
CAS Number:
Molecular Weight:
238.28
FEMA Number:
4703
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
09.780
NACRES:
NA.21
organoleptic:
balsamic
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

assay

≥98%

mp

31-33 °C

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

balsamic

SMILES string

O=C(OC/C=C/C1=CC=CC=C1)C2=CC=CC=C2

InChI

1S/C16H14O2/c17-16(15-11-5-2-6-12-15)18-13-7-10-14-8-3-1-4-9-14/h1-12H,13H2/b10-7+

InChI key

UARVBDPGNUHYQT-JXMROGBWSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service