Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W299620

Sigma-Aldrich

Rum ether

natural (US)

Synonym(s):

Ethyl oxyhydrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2996
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
organoleptic:
burnt; brown; ethereal; acidic; spicy; smoky; rum; whiskey
grade:
Fragrance grade
Kosher
natural (US)
agency:
follows IFRA guidelines
food allergen:
no known allergens

grade

Fragrance grade
Kosher
natural (US)

Quality Level

agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
FDA 21 CFR 117

refractive index

n20/D 1.366 (lit.)

bp

76 °C (lit.)

density

0.825 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

organoleptic

burnt; brown; ethereal; acidic; spicy; smoky; rum; whiskey

Looking for similar products? Visit Product Comparison Guide

Legal Information

For R&D use only.

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

75.0 °F

flash_point_c

23.9 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service