Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W271410

Sigma-Aldrich

α−Isomethyl ionone

≥95%, stabilized, FG

Synonym(s):

3-Methyl-4-(2,6,6-trimethyl-2-cyclohexen-1-yl)-3-buten-2-one, α-Cetone, Isomethyl-α-ionone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22O
CAS Number:
Molecular Weight:
206.32
FEMA Number:
2714
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
07.036
organoleptic:
floral; violet
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

assay

≥95%

contains

0.10% α-tocopherol, synthetic as stabilizer

density

0.93 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

organoleptic

floral; violet

SMILES string

CC(=O)C(\C)=C\C1C(C)=CCCC1(C)C

InChI

1S/C14H22O/c1-10-7-6-8-14(4,5)13(10)9-11(2)12(3)15/h7,9,13H,6,8H2,1-5H3/b11-9+

InChI key

JRJBVWJSTHECJK-PKNBQFBNSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

230.5 °F - closed cup

flash_point_c

110.3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service