Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P7171

Supelco

Pyrithioxin

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20N2O4S2
CAS Number:
Molecular Weight:
368.47
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

vitamins, nutraceuticals, and natural products

format

neat

SMILES string

Cc1ncc(CSSCc2cnc(C)c(O)c2CO)c(CO)c1O

InChI

1S/C16H20N2O4S2/c1-9-15(21)13(5-19)11(3-17-9)7-23-24-8-12-4-18-10(2)16(22)14(12)6-20/h3-4,19-22H,5-8H2,1-2H3

InChI key

SIXLXDIJGIWWFU-UHFFFAOYSA-N

General description

Pyrithioxin is a member of the vitamin B6 class of compounds.[1]

Application

Pyrithioxin may be used as an analytical reference standard for the determination of the analyte in pharmaceutical formulations and biological samples by spectrophotometric[2][3] and chromatographic[4][1] techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of Pyrithioxin and Its Metabolites in Blood and Urine of Dogs by High-Performance Liquid Chromatography
KITAO K, et al.
Chemical & Pharmaceutical Bulletin, 25(6), 1335-1342 (1977)
Spectrophotometric Determination of Pyrithioxin in Dosage Forms
Belal F, et al.
Analytical Letters, 19(13-14), 1533-1543 (1986)
Spectrophotometric determination of pyrithioxin in pharmaceutical preparations
Hassan SM, et al.
Spectroscopy Letters, 23(10), 1273-1284 (1990)
Ion-Pairing High-Performance Liquid Chromatographic Determintation of Pyridoxine and Pyrithioxine in Dosage Forms
Belal F
Analytical Letters, 22(8), 1897-1907 (1989)
Occupational contact dermatitis due to pyritinol.
W Wigger-Alberti et al.
Contact dermatitis, 37(2), 91-92 (1997-08-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service