Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M7384

Supelco

6-Methoxytryptamine

analytical standard

Synonym(s):

3-(2-Aminoethyl)-6-methoxyindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2O
CAS Number:
Molecular Weight:
190.24
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

form

crystalline

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

146-147 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc2c(CCN)c[nH]c2c1

InChI

1S/C11H14N2O/c1-14-9-2-3-10-8(4-5-12)7-13-11(10)6-9/h2-3,6-7,13H,4-5,12H2,1H3

InChI key

VOCGEKMEZOPDFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Liu et al.
Journal of medicinal chemistry, 50(1), 83-93 (2007-01-05)
Potassium ion (K+) channels are attractive targets for drug discovery because of the essential roles played in biological systems. However, high-throughput screening (HTS) cannot be used to screen K+ channel blockers. To overcome this disadvantage of HTS, we have developed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service