Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F1668

Sigma-Aldrich

Ammonium iron(III) sulfate dodecahydrate

BioXtra, ≥99%

Synonym(s):

Ammonium ferric sulfate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Fe(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
482.19
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:

product line

BioXtra

assay

≥99%

reaction suitability

core: iron

manufacturer/tradename

Cell Applications, Inc

impurities

≤0.0005% Phosphorus (P)
≤0.01% Insoluble matter

mp

39-41 °C (lit.)

solubility

dilute HCl: 1 M, clear, light brown

anion traces

chloride (Cl-): ≤0.001%

cation traces

Al: ≤0.01%
Ca: ≤0.001%
Cu: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

N.O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].OS([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Fe.H3N.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;1H3;2*(H2,1,2,3,4);12*1H2/q+3;;;;;;;;;;;;;;;/p-3

InChI key

LCPUDZUWZDSKMX-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Săndulescu et al.
Journal of pharmaceutical and biomedical analysis, 14(8-10), 951-958 (1996-06-01)
The method is based upon the reaction between fluoride ions and the coloured complex of Fe(III) with methyl salicylate to form the stable, colourless hexaflouride complex of iron. The conditions of the method (pH, time and combination ratio) were studied
S Puntarulo et al.
Archives of biochemistry and biophysics, 264(2), 482-491 (1988-08-01)
The interaction of microsomes with iron and NADPH to generate active oxygen radicals was determined by assaying for low level chemiluminescence. The ability of several ferric complexes to catalyze light emission was compared to their effect on microsomal lipid peroxidation
J G Reinhold et al.
The Journal of nutrition, 116(6), 1007-1017 (1986-06-01)
The effects of pH, ascorbate, citrate and dietary fiber on retention of ferrous and ferric iron by jejuno-ileal segments of rat intestine were examined in vivo. Iron was introduced in an isosmotic solution of sodium chloride and dextrose buffered by
I Wittmann et al.
Chemico-biological interactions, 138(2), 171-187 (2001-10-24)
Diabetes mellitus is characterized by increased methylglyoxal (MG) production. The aim of the present study was to investigate the role of iron in the cellular and molecular effects of MG. A red blood cell (RBC) model and L-arginine were used
Andrew J Ghio et al.
American journal of physiology. Lung cellular and molecular physiology, 283(3), L563-L572 (2002-08-10)
The iron chelator deferoxamine has been reported to inhibit both xanthine oxidase (XO) and xanthine dehydrogenase activity, but the relationship of this effect to the availability of iron in the cellular and tissue environment remains unexplored. XO and total xanthine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service