D3881
Diethylstilbestrol dipropionate
analytical standard, for drug analysis
Synonym(s):
(E)-3,4-Diphenyl-3-hexene p,p′-dipropionate, Stilbestrol dipropionate
About This Item
Recommended Products
grade
analytical standard, for drug analysis
Quality Level
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
forensics and toxicology
pharmaceutical (small molecule)
veterinary
format
neat
SMILES string
CCC(=O)Oc1ccc(cc1)\C(CC)=C(/CC)c2ccc(OC(=O)CC)cc2
InChI
1S/C24H28O4/c1-5-21(17-9-13-19(14-10-17)27-23(25)7-3)22(6-2)18-11-15-20(16-12-18)28-24(26)8-4/h9-16H,5-8H2,1-4H3/b22-21+
InChI key
VZMLEMYJUIIHNF-QURGRASLSA-N
General description
Application
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Repr. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service