Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

97582

Supelco

Acetazolamide

analytical standard

Synonym(s):

5-Acetamido-1,3,4-thiadiazole-2-sulfonamide, N-(5-Sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide, N-(5-[Aminosulfonyl]-1,3,4-thiadiazol-2-yl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N4O3S2
CAS Number:
Molecular Weight:
222.25
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

CC(=O)Nc1nnc(s1)S(N)(=O)=O

InChI

1S/C4H6N4O3S2/c1-2(9)6-3-7-8-4(12-3)13(5,10)11/h1H3,(H2,5,10,11)(H,6,7,9)

InChI key

BZKPWHYZMXOIDC-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Carbonic anhydrase inhibitor; increases cerebral blood flow.

Biochem/physiol Actions

Inhibits water permeability of membranes by interacting with aquaporins

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergio Caravita et al.
Chest, 147(1), 120-131 (2014-09-05)
Nocturnal periodic breathing occurs more frequently in men than in women with various clinical and pathophysiologic conditions. The mechanisms accounting for this sex-related difference are not completely understood. Acetazolamide effectively counteracts nocturnal periodic breathing, but it has been investigated almost
Sinan Bilginer et al.
Journal of enzyme inhibition and medicinal chemistry, 29(4), 495-499 (2013-07-03)
A series of phenols incorporating tertiary amine and trans-pyridylethenyl-carbonyl moieties were assayed as inhibitors of the β-carbonic anhydrase (CA, EC 4.2.1.1) from Saccharomyces cerevisiae, ScCA. One of these compounds was a low nanomolar ScCA inhibitor, whereas the remaining ones inhibited
Jyotsana Singh et al.
Drug development and industrial pharmacy, 40(9), 1223-1232 (2013-07-11)
The objective of research was to develop a novel pH-triggered polymeric nanoparticulate in situ gel (NP-ISG) for ophthalmic delivery of acetazolamide (ACZ) to enhance the conjunctival permeation and precorneal residence time of the formulation by overcoming the limitations of protective
Deborah S Schumacher et al.
Chest, 146(5), 1226-1236 (2014-07-06)
Sleep-disturbed breathing (SDB) is common in patients with precapillary pulmonary hypertension (PH). Nocturnal oxygen therapy (NOT) and acetazolamide improve SDB in patients with PH, and NOT improves exercise capacity. We investigated the effect of NOT and acetazolamide on nocturnal cardiac
Fatih Mehmet Türkcü et al.
Cutaneous and ocular toxicology, 33(2), 115-119 (2013-07-12)
The aim of this study was to evaluate the differences, in terms of visual outcome and treatment needs, between smokers and non-smokers central serous chorioretinopathy (CSCR) patients. The files of 252 patients diagnosed with CSCR who had presented to the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service