Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95353

Supelco

(+)-Dibenzyl L-tartrate

for chiral derivatization, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18O6
CAS Number:
Molecular Weight:
330.33
Beilstein/REAXYS Number:
3221733
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
NACRES:
NA.21

grade

for chiral derivatization

Quality Level

assay

≥98.0% (sum of enantiomers, HPLC)
≥98.0%

form

solid

optical activity

[α]20/D +12.5±1°, c = 1.1% in acetone

mp

51-54 °C

application(s)

general analytical

SMILES string

O[C@H]([C@@H](O)C(=O)OCc1ccccc1)C(=O)OCc2ccccc2

InChI

1S/C18H18O6/c19-15(17(21)23-11-13-7-3-1-4-8-13)16(20)18(22)24-12-14-9-5-2-6-10-14/h1-10,15-16,19-20H,11-12H2/t15-,16-/m1/s1

InChI key

LCKIPSGLXMCAOF-HZPDHXFCSA-N

Other Notes

Chiral auxiliary for preparing CAB catalysts [chiral (acyloxy)boranes]; Chiral selector for HPLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Furuta et al.
Organic Syntheses, 72, 86-86 (1993)
Y. Hu et al.
Journal of the American Chemical Society, 118, 4550-4550 (1996)
E. Heldin et al.
Chromatographia, 32, 407-407 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service