Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92656

Supelco

Benzyl formate

analytical standard

Synonym(s):

Benzyl methanoate, Formic acid benzyl ester, NSC 8049

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2CH2C6H5
CAS Number:
Molecular Weight:
136.15
Beilstein/REAXYS Number:
150-84-5
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.508-1.515
n20/D 1.511 (lit.)

bp

203 °C (lit.)

density

1.088 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

O=COCC1=CC=CC=C1

InChI

1S/C8H8O2/c9-7-10-6-8-4-2-1-3-5-8/h1-5,7H,6H2

InChI key

UYWQUFXKFGHYNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

179.6 °F - closed cup

flash_point_c

82 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service