Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92370

Sigma-Aldrich

1,1,3-Trimethylcyclohexane

purum, ≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18
CAS Number:
Molecular Weight:
126.24
Beilstein/REAXYS Number:
1900451
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

purum

assay

≥99.0% (GC)

refractive index

n20/D 1.429

bp

136-138 °C

density

0.778 g/mL at 20 °C (lit.)

SMILES string

CC1CCCC(C)(C)C1

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service