Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91844

Supelco

1-Bromopropane

analytical standard

Synonym(s):

Propyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2Br
CAS Number:
Molecular Weight:
122.99
Beilstein/REAXYS Number:
505936
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.3 (vs air)

vapor pressure

146 mmHg ( 20 °C)

assay

≥98.0% (GC)

autoignition temp.

914 °F

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.432-1.438
n20/D 1.434 (lit.)

bp

71 °C (lit.)

mp

−110 °C (lit.)

density

1.354 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CCCBr

InChI

1S/C3H7Br/c1-2-3-4/h2-3H2,1H3

InChI key

CYNYIHKIEHGYOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromopropane (1-BP) is widely used as a substitute for the ozone-depleting solvents. It is non-flammable and moderately volatile and known to exhibit neurotoxicity in both rodents and humans. 1-BP is used in vapor and immersion degreasing operations and is also used as a solvent in industries.[1][2]

Application

1-Bromopropane (1-BP) may be used as an analytical standard for the determination of the analyte in human urine by gas chromatography technique.[3][4]

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Ozone 1 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system, Liver,Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

76.1 °F - closed cup

flash_point_c

24.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Increased susceptibility of nrf2-null mice to 1-bromopropane--induced hepatotoxicity
Liu F, et al.
Toxicological Sciences, 115(2), 596-606 (2010)
Neurological disorders in three workers exposed to 1-bromopropane
Ichihara G, et al.
Journal of Occupational Health, 44(1), 1-7 (2002)
1-Bromopropane and 2-bromopropane--Determination of 1-bromopropane and 2-bromopropane in urine by dynamic headspace GC/MS [Biomonitoring Methods, 2019]
Ro?bach B, et al.
The MAK-Collection for Occupational Health and Safety, 4(1), 317-336 (2002)
Development of a headspace gas chromatographic test for the quantification of 1-and 2-bromopropane in human urine
B?Hymer C and Cheever KL
Journal of Chromatography. B, Biomedical Sciences and Applications, 814(1), 185-189 (2005)

Questions

  1. GC graph required for 1-Bromopropane, CAS No. 106-94-5 Product Number 91844, Batch No. BCCJ7550, Brand SIAL.

    1 answer
    1. The documentation you are requesting can be supplied by our Technical Service team who can assist you further. We kindly ask you to navigate to the link https://www.sigmaaldrich.com/techservice, and click on "Product Documentation" under the Products Section with all the required information so that a member of our team can reach out to you to assist further. Thank you.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service