Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91595

Supelco

Chamazulene

analytical standard

Synonym(s):

7-Ethyl-1,4-dimethylazulene, Chamazulen, Dimethulene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16
CAS Number:
Molecular Weight:
184.28
Beilstein/REAXYS Number:
1306577
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CCc1ccc(C)c2ccc(C)c2c1

InChI

1S/C14H16/c1-4-12-7-5-10(2)13-8-6-11(3)14(13)9-12/h5-9H,4H2,1-3H3

InChI key

GXGJIOMUZAGVEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: achillea

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

278.6 °F

flash_point_c

137 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Beáta Gosztola et al.
Natural product communications, 5(3), 465-470 (2010-04-28)
In our study we examined the variability of the essential oil content and composition of chamomile (Matricaria recutita L.) during three years (2005-2007). Twenty-eight populations of wild origin and 4 registered cultivars ('Soroksári 40', 'Lutea', 'Goral' and 'Bona') were evaluated
Saban Kordali et al.
Journal of agricultural and food chemistry, 53(5), 1408-1416 (2005-03-03)
The compositions of essential oils isolated from the aerial parts of Artemisia absinthium, Artemisia santonicum, and Artemisia spicigera by hydrodistillation were analyzed by GC-MS, and a total of 204 components were identified. The major components of these essential oils were
E A Rekka et al.
Research communications in molecular pathology and pharmacology, 92(3), 361-364 (1996-06-01)
Oxygen toxicity and related free radical reactions are implicated in numerous pathophysiological conditions, like atherosclerosis, inflammation, gastric ulceration, neuronal degeneration, tumour promotion. The flowers of Matricaria chamomilla, Asteraceae, have been used therapeutically for conditions in which oxidative stress is supposed
Mai Ramadan et al.
Journal of chromatography. A, 1133(1-2), 221-225 (2006-09-01)
A new, simple and sensitive high-performance liquid chromatographic (HPLC) method was developed for the determination of chamazulene carboxylic acid (CCA) in serum. The technique is based on a single liquid-liquid extraction of the substance using ibuprofen as internal standard (I.S.).
Anne Orav et al.
Natural product research, 20(12), 1082-1088 (2006-11-28)
Variations in the essential oil composition of Achillea millefolium L. growing in Estonia and in other European countries, were determined. The oils were obtained in yields of 0.9-9.5 mL kg-1. A total of 102 components were identified. The quantitatively most

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service