Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90985

Supelco

Diosmetin

analytical standard

Synonym(s):

3′,5,7-Trihydroxy-4′-methoxyflavone, 4′-Methylluteolin, 5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)chromen-4-one, Luteolin 4′-methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O6
CAS Number:
Molecular Weight:
300.26
Beilstein/REAXYS Number:
294492
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(cc1O)C2=CC(=O)c3c(O)cc(O)cc3O2

InChI

1S/C16H12O6/c1-21-13-3-2-8(4-10(13)18)14-7-12(20)16-11(19)5-9(17)6-15(16)22-14/h2-7,17-19H,1H3

InChI key

MBNGWHIJMBWFHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Flavonoid from Citrus limon. Antioxidant. Inhibits carcinogen activation by inhibiting the CYP1A1 enzyme.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marios Spanakis et al.
Biomedical chromatography : BMC, 23(2), 124-131 (2008-10-14)
Diosmetin and hesperetin are the aglycones of the flavonoid glycosides diosmin and hesperidin which occur naturally in citrus fruit. A GC/MS method for the simultaneous determination of diosmetin and hesperetin in human plasma and urine has been developed and validated.
Helana Naguib Michael et al.
Phytotherapy research : PTR, 27(5), 699-704 (2012-07-05)
The new natural flavonoid compounds - diosmetin 7-O-β-L-arabinofuranosyl (1 → 2) β-D-apiofuranoside (1) and diosmetin 7-O-β-D-apiofuranoside (2) - were isolated from the acetone extract of date fruits epicarp belonging to family Arecaceae (Palmae). Elucidation of their chemical structures was determined by different
Dave Chandler et al.
Molecular nutrition & food research, 54 Suppl 2, S141-S150 (2010-06-12)
Advanced glycation endproducts (AGEs) accumulate on protein deposits including the beta-amyloid plaques in Alzheimer's disease. AGEs interact with the "receptor for advanced glycation endproducts", and transmit their signals using intracellular reactive oxygen species as second messengers. Ultimately, AGEs induce the
Luigi Quintieri et al.
Biochemical pharmacology, 75(6), 1426-1437 (2008-01-15)
We evaluated the effects of increasing concentrations of the flavonoids salvigenin, diosmetin and luteolin on the in vitro metabolism of midazolam (MDZ), a probe substrate for cytochrome P450 (CYP) 3A enzymes, which is converted into 1'-hydroxy-midazolam (1'-OH-MDZ) and 4-hydroxy-midazolam (4-OH-MDZ)
Vasilis P Androutsopoulos et al.
The Journal of nutritional biochemistry, 24(2), 496-504 (2012-07-04)
Various types of tumors are known to overexpress enzymes belonging to the CYP1 family of cytochromes P450. The present study aimed to characterize the metabolism and further antiproliferative activity of the natural flavonoid diosmetin in the CYP1-expressing human hepatoma cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service