89985
γ-Undecalactone
analytical standard
Synonym(s):
Undecanoic γ-lactone, Aldehyde C14
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥98.5% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
refractive index
n20/D 1.45 (lit.)
bp
164-166 °C/13 mmHg (lit.)
density
0.949 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
neat
SMILES string
CCCCCCCC1CCC(=O)O1
InChI
1S/C11H20O2/c1-2-3-4-5-6-7-10-8-9-11(12)13-10/h10H,2-9H2,1H3
InChI key
PHXATPHONSXBIL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Gouda-type cheeses using gas chromatography coupled to mass spectrometry (GC-MS).[1]
- Strawberries using gas chromatography with flame ionization detection (GC-FID) and mass spectrometry (GC-MS).[2]
- Apricots (Prunus armeniaca) using high-resolution gas chromatography-mass spectrometry (HRGC-MS) and HRGC olfactometry (HRGC-O).[3]
hcodes
pcodes
Hazard Classifications
Aquatic Chronic 3
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
293.0 °F - closed cup
flash_point_c
145 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service