Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

83602

Sigma-Aldrich

Resorcinol

purum, ≥98.0% (HPLC)

Synonym(s):

1,3-Benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein/REAXYS Number:
906905
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor density

3.8 (vs air)

vapor pressure

1 mmHg ( 21.1 °C)

grade

purum

assay

≥98.0% (HPLC)

autoignition temp.

1126 °F

bp

178 °C/16 mmHg (lit.)

mp

109-112 °C (lit.)
109-112 °C

SMILES string

Oc1cccc(O)c1

InChI

1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H

InChI key

GHMLBKRAJCXXBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

may discolor on storage

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andriy I Vovk et al.
Bioorganic & medicinal chemistry letters, 19(5), 1314-1317 (2009-02-12)
Resorcinarene oxazines bearing four TEMPO fragments at the wide rim of the macrocycle were prepared through the aminomethylation of resorcinarene octols with 4-amino-TEMPO and formaldehyde. Tetra-TEMPO resorcinarenes are efficient scavengers of 1,1-diphenyl-2-picrylhydrazyl radicals. The model studies revealed that macrocyclic structure
S L da Silva et al.
European journal of medicinal chemistry, 44(1), 312-321 (2008-04-22)
Phospholipases A(2) are enzymes responsible for the hydrolysis of membrane phospholipids that release arachidonic acid, which serves as substrate for pro-inflammatory mediators, such as prostaglandins and leucotriens. The design of specific inhibitors for PLA(2) might help in the development of
Graziella Tocco et al.
Bioorganic & medicinal chemistry letters, 19(1), 36-39 (2008-12-05)
In this work, the PEG-immobilization and the liquid phase synthesis of some coumarins derived from cardol are presented. Some preliminary results on their tyrosinase inhibitory activity are also included.
Giovanni Appendino et al.
Journal of natural products, 71(8), 1427-1430 (2008-08-07)
Marijuana (Cannabis sativa) has long been known to contain antibacterial cannabinoids, whose potential to address antibiotic resistance has not yet been investigated. All five major cannabinoids (cannabidiol (1b), cannabichromene (2), cannabigerol (3b), Delta (9)-tetrahydrocannabinol (4b), and cannabinol (5)) showed potent
Alessio Innocenti et al.
Bioorganic & medicinal chemistry, 16(15), 7424-7428 (2008-06-27)
Inhibition of 12 mammalian isoforms of the metalloenzyme carbonic anhydrase (CA, EC 4.2.1.1), CA I-XIV, with a series of phenols was investigated. The inhibition profile by phenols of these CAs was distinct from those of the sulfonamides and their isosteres

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service