Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

83450

Sigma-Aldrich

Rapeseed oil from Brassica rapa

natural

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

form

oil

Quality Level

quality

natural

refractive index

n20/D 1.47

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Rapeseed oil from Brassica rapa is mainly used in cooking and also in salads.[1]

Storage Class

10 - Combustible liquids

wgk_germany

awg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mapping loci controlling the concentrations of erucic and linolenic acids in seed oil of Brassica napus L.
Thormann CE, et al.
Tag. Theoretical and Applied Genetics. Theoretische Und Angewandte Genetik, 93(1-2), 282-286 (1996)
Mingguo Tang et al.
The New phytologist, 196(2), 414-426 (2012-08-21)
Metabolic control analysis allows the study of metabolic regulation. We applied both single- and double-manipulation top-down control analysis to examine the control of lipid accumulation in developing oilseed rape (Brassica napus) embryos. The biosynthetic pathway was conceptually divided into two
Hernan Brice Kenmogne-Domguia et al.
Food & function, 3(12), 1302-1309 (2012-08-25)
During digestion, lipids undergo modifications of their colloidal and molecular structures, which depend on the digestive conditions and the composition of the digestive juices. The aim of this work was to evaluate whether gastric pH and pepsin modulate the colloidal
Jan Topinka et al.
Toxicology letters, 212(1), 11-17 (2012-05-09)
The present study was performed to identify possible genotoxicity induced by organic extracts from particulate matter in the exhaust of two typical diesel engines run on diesel fuel and neat heated fuel-grade rapeseed oil: a Cummins ISBe4 engine tested using
Andrea Baxheinrich et al.
The British journal of nutrition, 108(4), 682-691 (2012-08-17)
In therapy of the metabolic syndrome, the optimal dietary approach with regard to its macronutrient composition and metabolically favourable food components, such as the plant-derived n-3 fatty acid α-linolenic acid (ALA), is still a matter of debate. We investigated the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service