82580
Purine
puriss., ≥98.5% (HPLC)
Synonym(s):
7H-Imidazo(4,5-d)pyrimidine
About This Item
grade
puriss.
assay
≥98.5% (HPLC)
mp
214-217 °C (lit.)
215-217 °C
storage temp.
2-8°C
SMILES string
c1ncc2nc[nH]c2n1
InChI
1S/C5H4N4/c1-4-5(8-2-6-1)9-3-7-4/h1-3H,(H,6,7,8,9)
InChI key
KDCGOANMDULRCW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
replaced by
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service