Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

82551

Sigma-Aldrich

Pterine

≥97.0% (HPLC)

Synonym(s):

2-Amino-4-hydroxypteridine, 2-Amino-4-oxodihydropteridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5N5O
CAS Number:
Molecular Weight:
163.14
Beilstein/REAXYS Number:
150294
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

assay

≥97.0% (HPLC)

storage temp.

2-8°C

SMILES string

Nc1nc(O)c2nccnc2n1

InChI

1S/C6H5N5O/c7-6-10-4-3(5(12)11-6)8-1-2-9-4/h1-2H,(H3,7,9,10,11,12)

InChI key

HNXQXTQTPAJEJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Review: Biosynthesis and metabolism of tetrahydrobiopterin and molybdopterin[1]

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C l'Hélias et al.
Mutation research, 128(2), 147-152 (1984-09-01)
The experiments described in this paper show that synthetic pteridines, especially biopterin and pterin, injected directly into Drosophila melanogaster induce recessive lethals. On the contrary, D-neopterin seems to have little effect. A mutagenic effect has previously been shown for an
A S Lewis et al.
The Journal of biological chemistry, 259(1), 12-15 (1984-01-10)
We have examined the effects of folate compounds and the folate analog amethopterin (methotrexate) as inhibitors of mammalian xanthine oxidase and have found that they offer potent inhibition of the enzyme. We have compared the inhibitory potency of folic acid
Biosynthesis and metabolism of tetrahydrobiopterin and molybdopterin.
C A Nichol et al.
Annual review of biochemistry, 54, 729-764 (1985-01-01)
Matthew P Rubach et al.
PLoS pathogens, 11(3), e1004655-e1004655 (2015-03-13)
Decreased bioavailability of nitric oxide (NO) is a major contributor to the pathophysiology of severe falciparum malaria. Tetrahydrobiopterin (BH4) is an enzyme cofactor required for NO synthesis from L-arginine. We hypothesized that systemic levels of BH₄ would be decreased in
Daisuke Kadowaki et al.
Biological & pharmaceutical bulletin, 38(3), 487-492 (2015-03-12)
Uric acid exerts an important antioxidant effect against external oxidative stress under physiological conditions. However, uric acid itself can increase oxidative stress via reduced nicotinamide adenine dinucleotide phosphate (NADPH) oxidase activation in adipocytes and vascular cells. Uric acid transporter 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service