Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77522

Supelco

Ethyl vanillate

analytical standard

Synonym(s):

Ethyl 4-hydroxy-3-methoxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O4
CAS Number:
Molecular Weight:
196.20
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

Quality Level

grade

analytical standard

assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

O=C(OCC)C1=CC(OC)=C(O)C=C1

InChI

1S/C10H12O4/c1-3-14-10(12)7-4-5-8(11)9(6-7)13-2/h4-6,11H,3H2,1-2H3

InChI key

MWAYRGBWOVHDDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

252.0 °F

flash_point_c

122.2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosario Sánchez-Gómez et al.
Food chemistry, 329, 127181-127181 (2020-06-06)
The compounds that the wood releases to the wine and the oxygen transmission rate (OTR) of the barrel define the final wine. The new possibility of choosing the OTR of the barrel allows the winemaker to globally control the ageing
L Federico Casassa et al.
Food chemistry, 340, 127573-127573 (2020-10-09)
Cabernet Sauvignon wines were aged for 15 months in used and new French 225 L oak barrels, followed by a period of 3 months in bottle. In addition to control barrels (3 years old), three bending/toasting protocols, including fire bent and fire toasted
Hong-shik Lee et al.
Bioresource technology, 203, 142-149 (2016-01-02)
The hydro-/solvothermolysis of kraft lignin using water and ethanol as a solvent were investigated in this study. The effect of the water-to-ethanol ratio on the yields of monomeric aromatic chemicals (MACs) and the kinetic behavior of MACs was studied in
Josep Massana-Codina et al.
Frontiers in plant science, 11, 1287-1287 (2020-09-26)
Black dot is a blemish disease of potato tubers caused by the phytopathogenic fungus Colletotrichum coccodes. Qualitative resistance (monogenic) that leads to the hypersensitive response has not been reported against black dot, but commercial potato cultivars show different susceptibility levels
Fabiana Labanca et al.
Antioxidants (Basel, Switzerland), 9(8) (2020-08-08)
The leaves of Vitis vinifera L. have been used for a long time in traditional medicine for the treatment of many ailments. Grape polyphenols, indeed, have been demonstrated to be able to defend against oxidative stress, responsible for various disorders

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service