Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74192

Supelco

Picein

analytical standard

Synonym(s):

1-[4-(β-D-Glucopyranosyloxy)phenyl]ethan-1-one, 4′-(β-D-Glucopyranosyloxy)acetophenone, 4-Acetylphenyl β-D-glucopyranoside, p-Hydroxyacetophenone glucoside, Ameliaroside, Piceoside, Salicinerein, Salinigrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18O7
CAS Number:
Molecular Weight:
298.29
Beilstein/REAXYS Number:
90049
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

SMILES string

O[C@@H]1[C@@H](O)[C@H](OC2=CC=C(C(C)=O)C=C2)O[C@H](CO)[C@H]1O

InChI

1S/C14H18O7/c1-7(16)8-2-4-9(5-3-8)20-14-13(19)12(18)11(17)10(6-15)21-14/h2-5,10-15,17-19H,6H2,1H3/t10-,11-,12+,13-,14-/m1/s1

InChI key

GOZCEKPKECLKNO-RKQHYHRCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riitta Tegelberg et al.
Phytochemical analysis : PCA, 29(5), 493-499 (2018-03-10)
In ecological experiments, storage of plant material is often needed between harvesting and laboratory analyses when the number of samples is too large for immediate, fresh analyses. Thus, accuracy and comparability of the results call for pre-treatment methods where the
L B Lai et al.
Analytical biochemistry, 200(2), 365-369 (1992-02-01)
The neutral pH optimum beta-glucosidases of mammalian liver and almonds are each capable of hydrolyzing a number of plant glucosides, including L-picein (p-hydroxyacetophenone-beta-D-glucoside) and prunasin (D-mandelonitrile-beta-D-glucoside). Taking advantage of the marked differences in the spectra of the substrate/product pairs of
H Løkke
Ecotoxicology and environmental safety, 19(3), 301-309 (1990-06-01)
The concentrations of the glucoside picein and its aglucone piceol (4-hydroxy acetophenone) in the needles of Norway spruce (Picea abies (L.) Karsten) are considered indicators of plant stress. By use of two clones and a natural population of Norway spruce
K L LaMarco et al.
The Biochemical journal, 237(2), 469-476 (1986-07-15)
We have isolated from guinea-pig liver a broad-specificity beta-glucosidase of unknown function that utilizes as its substrate non-physiological aryl glycosides (e.g. 4-methylumbelliferyl beta-D-glucopyranoside, p-nitrophenyl beta-D-glucopyranoside). The present paper documents that this enzyme can be inhibited by various naturally occurring glycosides
Anu Lavola et al.
Phytochemistry, 153, 102-110 (2018-06-16)
The phenolic phytochemicals of winter-dormant Salix pyrolifolia were determined from the vegetative buds, and the bark and wood of different-aged twigs by HPLC-DAD and UHPLC-QTOF-MS analyses. All the plant parts were composed of salicylate glucosides and the other Salix-specific, simple

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service