73962
I3,II8-Biapigenin
analytical standard
Synonym(s):
3,8″-Biapigenin, 5,5′,7,7′-Tetrahydroxy-2,2′-bis(4-hydroxyphenyl)-[3,8′-bi-4H-1-benzopyran]-4,4′-dione
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥98.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
impurities
≤10.0% water
application(s)
food and beverages
format
neat
storage temp.
2-8°C
SMILES string
Oc1ccc(cc1)C2=CC(=O)c3c(O)cc(O)c(c3O2)C4=C(Oc5cc(O)cc(O)c5C4=O)c6ccc(O)cc6
InChI
1S/C30H18O10/c31-15-5-1-13(2-6-15)22-12-21(37)24-19(35)11-20(36)26(30(24)39-22)27-28(38)25-18(34)9-17(33)10-23(25)40-29(27)14-3-7-16(32)8-4-14/h1-12,31-36H
InChI key
IQAMTZLKUHMPPE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service