73422
Sakuranetin
analytical standard
Synonym(s):
4′-5-Dihydroxy-7-methoxyflavanone, 7-O-Methylnaringenin, Naringenin 7-O-methyl ether
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥98.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
impurities
≤5.0% water
application(s)
food and beverages
format
neat
SMILES string
COc1cc(O)c2C(=O)C[C@H](Oc2c1)c3ccc(O)cc3
InChI
1S/C16H14O5/c1-20-11-6-12(18)16-13(19)8-14(21-15(16)7-11)9-2-4-10(17)5-3-9/h2-7,14,17-18H,8H2,1H3/t14-/m0/s1
InChI key
DJOJDHGQRNZXQQ-AWEZNQCLSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Aerial parts of Rhus retinorrhoea and Rhus tripartita by chloroform and methanol extractions followed by high performance thin-layer chromatography (HPTLC).[3]
- Rhamnus davurica by high performance liquid chromatography (HPLC) coupled to tandem mass spectrometry with electrospray ionization (ESI-MS/MS)[4] as well as affinity ultrafiltration followed by analysis using ESI-HPLC-MS/MS.[5]
- Royal jelly nutraceutical products by turbulent flow chromatography (TFC) combined with LC-Extractive-Orbitrap analyzer equipped with ESI.[1]
Packaging
Other Notes
Recommended products
signalword
Warning
hcodes
pcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service