Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72644

Sigma-Aldrich

Pyruvic acid

technical, ≥95.0% (T)

Synonym(s):

α-Ketopropionic acid, 2-Oxopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCOOH
CAS Number:
Molecular Weight:
88.06
Beilstein/REAXYS Number:
506211
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

technical

assay

≥95.0% (T)

form

liquid

refractive index

n20/D 1.427-1.434
n20/D 1.428 (lit.)

bp

165 °C (lit.)

mp

11-12 °C (lit.)

density

1.272 g/mL at 20 °C
1.267 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)C(O)=O

InChI

1S/C3H4O3/c1-2(4)3(5)6/h1H3,(H,5,6)

InChI key

LCTONWCANYUPML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

183.2 °F - closed cup

flash_point_c

84 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert A Jacobs et al.
American journal of physiology. Endocrinology and metabolism, 304(7), E686-E694 (2013-02-07)
Lactate is an important intermediate metabolite in human bioenergetics and is oxidized in many different tissues including the heart, brain, kidney, adipose tissue, liver, and skeletal muscle. The mechanism(s) explaining the metabolism of lactate in these tissues, however, remains unclear.
Hannes Link et al.
Nature biotechnology, 31(4), 357-361 (2013-03-05)
Recent data suggest that the majority of proteins bind specific metabolites and that such interactions are relevant to metabolic and gene regulation. However, there are no methods to systematically identify functional allosteric protein-metabolite interactions. Here we present an experimental and
Sarah C Atkinson et al.
Plant molecular biology, 81(4-5), 431-446 (2013-01-29)
Lysine is one of the most limiting amino acids in plants and its biosynthesis is carefully regulated through inhibition of the first committed step in the pathway catalyzed by dihydrodipicolinate synthase (DHDPS). This is mediated via a feedback mechanism involving
Emilie A Bard-Chapeau et al.
Nature genetics, 46(1), 24-32 (2013-12-10)
The most common risk factor for developing hepatocellular carcinoma (HCC) is chronic infection with hepatitis B virus (HBV). To better understand the evolutionary forces driving HCC, we performed a near-saturating transposon mutagenesis screen in a mouse HBV model of HCC.
Ching-An Wu et al.
Biochimica et biophysica acta, 1833(5), 1147-1156 (2013-02-05)
The Warburg effect is known to be crucial for cancer cells to acquire energy. Nutrient deficiencies are an important phenomenon in solid tumors, but the effect on cancer cell metabolism is not yet clear. In this study, we demonstrate that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service