Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

71541

Sigma-Aldrich

Sodium formate

puriss. p.a., ACS reagent, ≥99.0% (NT)

Synonym(s):

Formic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOONa
CAS Number:
Molecular Weight:
68.01
Beilstein/REAXYS Number:
3595134
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
39021901
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

assay

≥99.0% (NT)

pH

7.0-8.5 (25 °C, 50 mg/mL in H2O)

mp

259-262 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤100 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Na+].[O-]C=O

InChI

1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1

InChI key

HLBBKKJFGFRGMU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica Momb et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(2), 549-554 (2012-12-26)
Maternal supplementation with folic acid is known to reduce the incidence of neural tube defects (NTDs) by as much as 70%. Despite the strong clinical link between folate and NTDs, the biochemical mechanisms through which folic acid acts during neural
Sheng Feng et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 915-916, 28-32 (2013-01-15)
This paper described a sensitive and rapid method based on ultra-performance liquid chromatography coupled to tandem mass spectrometry (UPLC-MS/MS) for the determination of phenylephrine in human plasma. Plasma samples were pre-purified by solid-phase extraction (SPE). The chromatographic separation was achieved
Kyle C Costa et al.
mBio, 4(2), doi:10-doi:10 (2013-02-28)
Hydrogenotrophic methanogenic Archaea require reduced ferredoxin as an anaplerotic source of electrons for methanogenesis. H(2) oxidation by the hydrogenase Eha provides these electrons, consistent with an H(2) requirement for growth. Here we report the identification of alternative pathways of ferredoxin
Scott D Hamilton-Brehm et al.
Extremophiles : life under extreme conditions, 17(2), 251-263 (2013-01-25)
A novel sulfate-reducing bacterium designated OPF15(T) was isolated from Obsidian Pool, Yellowstone National Park, Wyoming. The phylogeny of 16S rRNA and functional genes (dsrAB) placed the organism within the family Thermodesulfobacteriaceae. The organism displayed hyperthermophilic temperature requirements for growth with
Cláudia Freitas et al.
Applied biochemistry and biotechnology, 169(7), 2038-2048 (2013-01-30)
The use of lignocellulosic materials as substrate for bioethanol production is considered a cost-effective approach to make the biofuel production process economically sustainable. However, lignocellulosic hydrolysis releases toxic compounds such as weak acids which inhibit microorganism growth and ethanol production.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service