Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

71193

Sigma-Aldrich

Sodium acetate trihydrate

purum p.a., ≥99.0% (NT)

Synonym(s):

Acetic acid sodium salt trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
Beilstein/REAXYS Number:
3732037
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

purum p.a.

assay

≥99.0% (NT)

autoignition temp.

1112 °F

pH

7.5-9.0 (25 °C, 50 mg/mL in H2O)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.O.O.[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na.3H2O/c1-2(3)4;;;;/h1H3,(H,3,4);;3*1H2/q;+1;;;/p-1

InChI key

AYRVGWHSXIMRAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium acetate trihydrate can be obtained from the crystallization of sodium acetate in water. It loses water of crystallization on heating at 75°C, while on heating above 120°C, anhydrous sodium acetate is obtained. Its aqueous solutions are basic in nature. ESR spectra of its irradiated single crystals have been studied at 77°K. The hyperfine proton and 13C splittings indicated the presence of trapped methyl radicals. Its crystal structure has been examined.

application

Sodium acetate trihydrate may be used in the preparation of silicone/sodium acetate trihydrate hybrid.

Other Notes

Enzyme substrate for acetate kinase (EC 2.7.2.1)

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal structure of sodium acetate trihydrate.
Cameron TS, et al.
Acta Crystallographica Section B, Structural Science, 32(1), 87-90 (1976)
Eagleson M.
Concise Encyclopedia Chemistry, 995-995 (1994)
Methyl radical in irradiated single crystals of sodium acetate trihydrate.
Rogers MT and Kispert LD.
J. Chem. Phys. , 46(1), 221-223 (1967)
Water-responsive shape memory hybrid: Design concept and demonstration.
Fan K, et al.
Express Polymer Letters, 5(5), 409-416 (2011)
Andrew D Shaw et al.
Annals of surgery, 255(5), 821-829 (2012-04-04)
To assess the association of 0.9% saline use versus a calcium-free physiologically balanced crystalloid solution with major morbidity and clinical resource use after abdominal surgery. 0.9% saline, which results in a hyperchloremic acidosis after infusion, is frequently used to replace

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service