Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68812

Supelco

Isobutyraldehyde diethyl acetal

analytical standard

Synonym(s):

1,1-Diethoxy-2-methylpropane, 1,1-Diethoxyisobutane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18O2
CAS Number:
Molecular Weight:
146.23
Beilstein/REAXYS Number:
1697899
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.394

density

0.823 g/mL at 20 °C

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

InChI

1S/C8H18O2/c1-5-9-8(7(3)4)10-6-2/h7-8H,5-6H2,1-4H3

InChI key

KZDFOVZPOBSHDH-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

73.2 °F

flash_point_c

22.9 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service