67690
Iodomethane
puriss., redist., ≥99.5% (GC)
Synonym(s):
Methyl iodide
About This Item
Recommended Products
vapor density
4.89 (vs air)
vapor pressure
24.09 psi ( 55 °C)
7.89 psi ( 20 °C)
grade
puriss.
assay
≥99.5% (GC)
quality
redist.
contains
silver wool as stabilizer
refractive index
n20/D 1.530
n20/D 1.531 (lit.)
bp
41-43 °C (lit.)
mp
−64 (lit.)
solubility
water: soluble 14 g/L at 20 °C
density
2.28 g/mL at 25 °C (lit.)
SMILES string
CI
InChI
1S/CH3I/c1-2/h1H3
InChI key
INQOMBQAUSQDDS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
replaced by
signalword
Danger
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
89.8 °F - closed cup
flash_point_c
32.1 °C - closed cup
ppe
Faceshields, Gloves, Goggles
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service