Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

64100

Sigma-Aldrich

Methacrylic anhydride

technical, ≥92% (T)

Synonym(s):

Dimethacrylic acid anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=C(CH3)CO]2O
CAS Number:
Molecular Weight:
154.16
Beilstein/REAXYS Number:
1761982
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

technical

assay

≥92% (T)

contains

~0.2% topanol A as stabilizer

refractive index

n20/D 1.453 (lit.)
n20/D 1.453

bp

87 °C/13 mmHg (lit.)

density

1.04 g/mL at 20 °C
1.035 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OC(=O)C(C)=C

InChI

1S/C8H10O3/c1-5(2)7(9)11-8(10)6(3)4/h1,3H2,2,4H3

InChI key

DCUFMVPCXCSVNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Hwa Kim et al.
Acta biomaterialia, 12, 21-29 (2014-12-03)
Degradation of the nucleus pulposus (NP) is an early hallmark of intervertebral disc degeneration. The capacity for endogenous regeneration in the NP is limited due to the low cellularity and poor nutrient and vascular supply. Towards restoring the NP, a
Wenjuan Liu et al.
Materials science & engineering. C, Materials for biological applications, 110, 110670-110670 (2020-03-25)
Periosteum as an important component in the construct of bone is mainly responsible for providing nourishment and regulating osteogenic differentiation. When bone defect happens, the functionality of periosteum will also be influenced, furthermore, it will finally hamper the process of
Weili Ma et al.
Biomolecules, 9(10) (2019-09-25)
Hyaluronic acid (HA) is a polysaccharide polymer frequently used as a starting material to fabricate hydrogels, especially for recapitulating the brain's extracellular matrix (ECM) for in vitro neural stem cell (NSC) cultures. Here, we report the successful synthesis of a
Praveen Bandaru et al.
Small (Weinheim an der Bergstrasse, Germany), 16(25), e2001837-e2001837 (2020-05-19)
Stem cells secrete trophic factors that induce angiogenesis. These soluble factors are promising candidates for stem cell-based therapies, especially for cardiovascular diseases. Mechanical stimuli and biophysical factors presented in the stem cell microenvironment play important roles in guiding their behaviors.
Zhao Wei et al.
Cell stem cell, 27(5), 798-812 (2020-09-16)
Matrix dynamics influence how individual cells develop into complex multicellular tissues. Here, we develop hydrogels with identical polymer components but different crosslinking capacities to enable the investigation of mechanisms underlying vascular morphogenesis. We show that dynamic (D) hydrogels increase the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service