Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

60160

Sigma-Aldrich

Potassium cyanate

purum, crystallized, ≥97.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOCN
CAS Number:
Molecular Weight:
81.12
Beilstein/REAXYS Number:
3594798
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

purum

assay

≥97.0% (AT)

quality

crystallized

density

2.056 g/mL at 25 °C (lit.)

SMILES string

[K+].[O-]C#N

InChI

1S/CHNO.K/c2-1-3;/h3H;/q;+1/p-1

InChI key

GKKCIDNWFBPDBW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 289-289 (1994)
Transformation of aldoses into glycosylamine 1, 2-(cyclic carbamates)(glyco-oxazolidin-2-ones) by reaction with potassium cyanate.
Kovacs J, et al.
Carbohydrate Research, 210, 155-166 (1991)
Eagleson M.
Concise Encyclopedia Chemistry, 503-503 (1994)
Copper-Catalyzed Coupling of Arylboronic Acids with Potassium Cyanate: A New Approach to the Synthesis of Aryl Carbamates.
Kianmehr E and Baghersad MH.
Advanced Synthesis & Catalysis, 353(14-15), 2599-2603 (2011)
Jeroen N Stoop et al.
PloS one, 9(7), e102163-e102163 (2014-07-16)
The immune response to post-translationally modified antigens is a key characteristic of rheumatoid arthritis. Carbamylation is such a posttranslational modification. Recently, we demonstrated that autoantibodies recognizing carbamylated proteins are present in sera of rheumatoid arthritis. The molecular mechanisms underlying the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service