Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59780

Supelco

DL-Kavain

analytical standard

Synonym(s):

trans-5,6-Dihydro-4-methoxy-6-(2-phenylethenyl)-2H-pyran-2-one, DL-Kawain

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14O3
CAS Number:
Molecular Weight:
230.26
Beilstein/REAXYS Number:
177877
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

142-148 °C

application(s)

food and beverages

format

neat

SMILES string

COC1=CC(=O)OC(C1)\C=C\c2ccccc2

InChI

1S/C14H14O3/c1-16-13-9-12(17-14(15)10-13)8-7-11-5-3-2-4-6-11/h2-8,10,12H,9H2,1H3/b8-7+

InChI key

XEAQIWGXBXCYFX-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DL-Kavain, an analog of yangonin, is a kavalactone found in the kava plant.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James M Mathews et al.
Drug metabolism and disposition: the biological fate of chemicals, 33(10), 1555-1563 (2005-07-22)
Reported adverse drug interactions with the popular herb kava have spurred investigation of the mechanisms by which kava could mediate these effects. In vivo and in vitro experiments were conducted to examine the effects of kava extract and individual kavalactones
M Villain et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 798(2), 351-354 (2003-12-04)
A sensitive, specific and reproducible method for the quantitative determination of kavain in human hair has been developed. The sample preparation involved a decontamination step of the hair with methylene chloride. The hair sample (about 50 mg) was incubated in
Huaiping Yuan et al.
Journal of clinical periodontology, 38(11), 1029-1036 (2011-11-19)
The purpose of this study was to assess the role of anti-bone resorptive agents and an anti-inflammatory compound in murine Porphyromonas gingivalis (P. gingivalis)-induced periodontitis. Six randomly assigned groups were administered vehicle (saline, control) (n = 6), P. gingivalis infection only (untreated)
Roberto Cirilli et al.
Journal of separation science, 31(12), 2206-2210 (2008-06-20)
First baseline HPLC enantioseparation of kavain is described. Complete enantiodiscrimination was achieved on the immobilised-type Chiralpak IA chiral stationary phase (CSP) using pure methanol and simple methanol-water and ethanol-water mixtures as eluents. A water-dependent enantioselectivity was clearly demonstrated. Performance of
Shuang Fu et al.
Biomedical chromatography : BMC, 23(1), 81-91 (2008-07-29)
Three novel, simple and reproducible high-performance liquid chromatography quantitative assays with UV detection were developed and validated for three major kavalactones--desmethoxyyangonin, methysticin and kavain--in rat liver microsomes using diazepam as an internal standard; liquid-liquid extraction was used for sample preparation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service