Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58498

Supelco

Isobutylbenzene

analytical standard

Synonym(s):

2-Methyl-1-phenylpropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(CH3)2
CAS Number:
Molecular Weight:
134.22
Beilstein/REAXYS Number:
1852218
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

4.2 mmHg ( 37.7 °C)

assay

≥99.5% (GC)

autoignition temp.

802 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.486 (lit.)
n20/D 1.486

bp

170 °C/736 mmHg (lit.)

mp

−51 °C (lit.)

density

0.853 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CC(C)Cc1ccccc1

InChI

1S/C10H14/c1-9(2)8-10-6-4-3-5-7-10/h3-7,9H,8H2,1-2H3

InChI key

KXUHSQYYJYAXGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

FlameEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

118.6 °F - closed cup

flash_point_c

48.1 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan P Graves et al.
Journal of medicinal chemistry, 48(11), 3714-3728 (2005-05-27)
Molecular docking is widely used to predict novel lead compounds for drug discovery. Success depends on the quality of the docking scoring function, among other factors. An imperfect scoring function can mislead by predicting incorrect ligand geometries or by selecting
Brian D Fitz et al.
Journal of chromatography. A, 1392, 82-90 (2015-03-31)
Low thermal mass gas chromatography (LTM-GC) was evaluated for rapid, high peak capacity separations with three injection methods: liquid, headspace solid phase micro-extraction (HS-SPME), and direct vapor. An Agilent LTM equipped with a short microbore capillary column was operated at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service