Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58261

Supelco

Hexyl valerate

puriss. p.a., standard for GC, ≥99.7% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)2CH2COO(CH2)5CH3
CAS Number:
Molecular Weight:
186.29
Beilstein/REAXYS Number:
1760230
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

standard for GC
puriss. p.a.

assay

≥99.7% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.421

density

0.863 g/mL at 20 °C (lit.)

application(s)

environmental

SMILES string

CCCCCCOC(=O)CCCC


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service