52210
Hexadecane
purum, ≥98.0% (GC)
Synonym(s):
n-Hexadecane, Cetane
About This Item
Recommended Products
vapor density
7.8 (vs air)
vapor pressure
1 mmHg ( 105.3 °C)
grade
purum
assay
≥98.0% (GC)
autoignition temp.
395 °F
impurities
olefins, none detected
refractive index
n20/D 1.434 (lit.)
n20/D 1.435
bp
287 °C (lit.)
mp
18 °C (lit.)
transition temp
solidification point 17.5-18.5 °C
density
0.773 g/mL at 25 °C (lit.)
SMILES string
CCCCCCCCCCCCCCCC
InChI
1S/C16H34/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3-16H2,1-2H3
InChI key
DCAYPVUWAIABOU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Replaced by
signalword
Danger
hcodes
Hazard Classifications
Asp. Tox. 1
supp_hazards
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
233.6 °F
flash_point_c
112 °C
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service