Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52007

Supelco

(+)-Dihydromethysticin

analytical standard

Synonym(s):

(6S)-6-[2-(1,3-Benzodioxol-5-yl)ethyl]-5,6-dihydro-4-methoxy-2H-pyran-2-one, Dihydromethysticin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O5
CAS Number:
Molecular Weight:
276.28
Beilstein/REAXYS Number:
91699
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COC1=CC(=O)O[C@@H](CCc2ccc3OCOc3c2)C1

InChI

1S/C15H16O5/c1-17-12-7-11(20-15(16)8-12)4-2-10-3-5-13-14(6-10)19-9-18-13/h3,5-6,8,11H,2,4,7,9H2,1H3/t11-/m0/s1

InChI key

RSIWXFIBHXYNFM-NSHDSACASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Greg E Petersen et al.
Journal of ethnopharmacology, 235, 301-308 (2019-02-03)
Kava and kava extracts have shown great potential as a way to minimize anxiety-associated symptoms and to help alleviate pain. Hepatoxicity has been associated with the consumption of kava products. The chemical compounds, kavalactones (KL) and flavokavains (FK) have been
Yijin Tang et al.
Molecules (Basel, Switzerland), 24(7) (2019-04-03)
An ultra-high-performance liquid chromatographic (UHPLC) separation was developed for six kava pyrones (methysticin, dihydromethysticin (DHM), kavain, dihydrokavain (DHK), desmethoxyyangonin (DMY), and yangonin), two unidentified components, and three Flavokavains (Flavokavain A, B, and C) in Piper methysticum (kava). The six major

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service