Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48754

Sigma-Aldrich

Genistein 4′,7-dimethyl ether

~95% (TLC)

Synonym(s):

4′,7-Dimethoxy-5-hydroxyisoflavone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O5
CAS Number:
Molecular Weight:
298.29
Beilstein/REAXYS Number:
296689
MDL number:
UNSPSC Code:
12352200

assay

~95% (TLC)

SMILES string

COc1ccc(cc1)C2=COc3cc(OC)cc(O)c3C2=O

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service