Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46518

Supelco

Nitromide

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3CONH2
CAS Number:
Molecular Weight:
211.13
Beilstein/REAXYS Number:
7096825
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

183-185 °C (lit.)

application(s)

clinical
forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

NC(=O)c1cc(cc(c1)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C7H5N3O5/c8-7(11)4-1-5(9(12)13)3-6(2-4)10(14)15/h1-3H,(H2,8,11)

InChI key

UUKWKUSGGZNXGA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N W Shappell et al.
Chemosphere, 38(8), 1757-1762 (1999-04-02)
Colostomized chickens given oral doses of 3,5-dinitrobenzamide (nitromide) cleared nitromide predominantly through the urine (58% of dose) and feces (21% of dose). Rats cleared 52% of nitromide via urinary excretion and 44% via feces. Major urinary metabolites for both chickens
V Facchini et al.
Xenobiotica; the fate of foreign compounds in biological systems, 10(4), 299-305 (1980-04-01)
1. Nitromide (3,5-dinitrobenzamide) is reduced to monoamino- and diamino-metabolites in vitro on anaerobic incubation with rat intestinal microflora. This conversion is suppressed by the antibiotics neomycin, tetracycline and bacitracin. 2. Nitromide is also in part reduced to monoamino but not
Núria Rubio et al.
Journal of chromatography. A, 1217(8), 1183-1190 (2009-12-22)
The racemic compound (+/-)-N-(3,4-cis-3-decyl-1,2,3,4-tetrahydrophenanthren-4-yl)-3,5-dinitrobenzamide ((+/-)-1), an analogue of increased lipophilicity of the chiral selector (CS) contained in the Whelk-O HPLC chiral stationary phase (CSP) has been resolved into its enantiomers by applying centrifugal partition chromatography (CPC). Considering the known enantioselectivity
A A Ensafi et al.
Colloids and surfaces. B, Biointerfaces, 87(2), 480-488 (2011-07-06)
A new catechol-derivative compound, N-(3,4-dihydroxyphenethyl)-3,5-dinitrobenzamide, was synthesized and used to construct a modified-carbon nanotubes paste electrode. The electro-oxidation of captopril at the surface of the modified electrode was studied using cyclic voltammetry, chronoamperometry, and electrochemical impedance spectroscopy. Under the optimized
V Facchini et al.
Xenobiotica; the fate of foreign compounds in biological systems, 10(4), 289-297 (1980-04-01)
1. Following oral administration of nitromide (3,5-dinitrobenzamide) to rats, 67.9% of the dose was excreted in urine and 32.6% in the faeces in 96 h. Significant biliary excretion of nitromide metabolites also occurred, although no evidence of enterohepatic cycling was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service