Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45492

Supelco

Fentin chloride

PESTANAL®, analytical standard

Synonym(s):

Triphenyltin chloride, Chlorotriphenylstannane, Fentin chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3SnCl
CAS Number:
Molecular Weight:
385.47
Beilstein/REAXYS Number:
524762
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

240 °C/13.5 mmHg (lit.)

mp

108 °C (dec.) (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

Cl[Sn](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/3C6H5.ClH.Sn/c3*1-2-4-6-5-3-1;;/h3*1-5H;1H;/q;;;;+1/p-1

InChI key

NJVOZLGKTAPUTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

>302.0 °F

flash_point_c

> 150 °C

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Antonio Ortiz et al.
Biochimica et biophysica acta, 1720(1-2), 137-142 (2006-02-14)
Organotin compounds are widely distributed toxicants. They are membrane-active molecules with broad biological toxicity. In this contribution, we study the effect of triorganotin compounds on membrane permeability using phospholipid model membranes and human erythrocytes. Tribultyltin and triphenyltin are able to
Yoshikazu Miura et al.
Toxicology, 299(2-3), 165-171 (2012-06-06)
Oral administration of triphenyltin chloride (TPT) (6 mg/100g body weight) inhibits insulin secretion by decreasing glucose-induced cytoplasmic Ca(2+) concentration ([Ca(2+)](i)) in pancreatic β-cells of the hamster. To test the possibility that the abnormal level of the [Ca(2+)](i) induced by TPT
Toshihiro Horiguchi et al.
Analytical and bioanalytical chemistry, 396(2), 597-607 (2009-11-03)
To examine the role of the retinoid X receptor (RXR) in the development of imposex in gastropods, we investigated the time course of expression of the RXR gene in various tissues (ctenidium, ovary or testis, digestive gland, penis-forming area or
G Janer et al.
The Journal of steroid biochemistry and molecular biology, 99(2-3), 147-156 (2006-04-20)
In a recent study, we demonstrated that androstenedione was mainly converted to testosterone (T) and 5alpha-dihydrotestosterone (DHT) by digestive gland/gonad complex microsomal fractions isolated from male Marisa cornuarietis, whereas it was primarily metabolized to 5alpha-dihydroandrostenedione (DHA) by females. In the
Bao-an Wang et al.
Zhonghua nan ke xue = National journal of andrology, 12(6), 516-519 (2006-07-13)
To investigate the effects of tributyltin chloride (TBT) and triphenyltin chloride (TPT) on rat testicular Leydig cells. The rat Leydig cells (LC-540) were incubated with 0 to 80 nmol/L TBT and TPT for 24 to approximately 96 h, and then

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service