Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45378

Supelco

Chlordane (technical mixture)

PESTANAL®, analytical standard

Synonym(s):

Chlordane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6Cl8
CAS Number:
Molecular Weight:
409.78
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

ClC1CC2C(C1Cl)[C@@]3(Cl)C(Cl)=C(Cl)[C@]2(Cl)C3(Cl)Cl

InChI

1S/C10H6Cl8/c11-3-1-2-4(5(3)12)9(16)7(14)6(13)8(2,15)10(9,17)18/h2-5H,1H2/t2?,3?,4?,5?,8-,9+/m0/s1

InChI key

BIWJNBZANLAXMG-YQELWRJZSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martina Koblizkova et al.
Environmental science & technology, 46(1), 391-396 (2011-11-23)
As part of continued efforts under the Global Atmospheric Passive Sampling (GAPS) Network to develop passive air samplers applicable to a wide-range of compounds, sorbent-impregnated polyurethane foam (SIP) disk samplers were codeployed and tested against conventional polyurethane foam (PUF) disk
Xianlei Zhu et al.
Journal of environmental monitoring : JEM, 14(7), 1926-1934 (2012-06-02)
Measurements of semi-volatile organic compounds (SVOCs) in air are subject to substantial variability and uncertainty. This study apportions total variance of polycyclic aromatic hydrocarbons (PAHs) and chlordanes to variability and uncertainty components. Concentrations of PAHs and chlordanes were measured inside
Pengfei Xiao et al.
New biotechnology, 29(1), 107-115 (2011-07-02)
There is very limited information on the biotransformation of organochlorine pesticide chlordane by microorganisms, and no systematic study on the metabolic products and pathways for chlordane transformation by wood-rot fungi has been conducted. In this study, trans-chlordane was metabolized with
Andres Martinez et al.
Environmental pollution (Barking, Essex : 1987), 161, 222-228 (2012-01-11)
Residential soils from Cedar Rapids, Iowa, USA were collected and analyzed for chlordanes and polychlorinated biphenyls (PCBs). This study is one of the very few urban soil investigations in the USA. The chlordanes concentrations ranged from 0 to 7500 ng
Marina Albentosa et al.
The Science of the total environment, 435-436, 430-445 (2012-08-14)
SFG and physiological rates were measured in wild mussels from the Spanish Marine Pollution monitoring program (SMP) in order to determine seawater quality. It consists of 41 stations, covering almost 2500 km of coast, making the SMP the widest-ranging monitoring

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service