43966
Thiram
certified reference material, TraceCERT®
Synonym(s):
Tetramethylthiuram disulfide, Bis(dimethylthiocarbamoyl) disulfide, Bis(dimethylthiocarbamyl) disulfide, Thiram
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
autoignition temp.
316 °F
shelf life
limited shelf life, expiry date on the label
storage condition
under inert gas (argon)
technique(s)
gas chromatography (GC): suitable
mp
156-158 °C (lit.)
format
neat
SMILES string
CN(C)C(=S)SSC(=S)N(C)C
InChI
1S/C6H12N2S4/c1-7(2)5(9)11-12-6(10)8(3)4/h1-4H3
InChI key
KUAZQDVKQLNFPE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
Legal Information
signalword
Warning
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral
target_organs
Liver
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service