Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43778

Supelco

Carvacrol methyl ether

analytical standard

Synonym(s):

2-Methoxy-p-cymol, 4-Isopropyl-2-methoxytoluene, 5-Isopropyl-2-methylanisole, Methylcarvacrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16O
CAS Number:
Molecular Weight:
164.24
Beilstein/REAXYS Number:
2412706
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

0.936 g/mL at 20 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CC1=C(OC)C=C(C(C)C)C=C1

InChI

1S/C11H16O/c1-8(2)10-6-5-9(3)11(7-10)12-4/h5-8H,1-4H3

InChI key

YVLHTQPPMZOCOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

167.0 °F - closed cup

flash_point_c

75 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongpeng Ma et al.
Journal of ethnopharmacology, 231, 386-393 (2018-11-25)
A folk herb Adenosma buchneroides found in the previous ethnobotanical investigation plays an important role as an insect repellent among the Aini people in southwest of China, but the active compounds responsible for repellent activity of the plant have not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service