Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42924

Supelco

Germacrone

analytical standard

Synonym(s):

(E,E)-Germacra-3,7(11),9-trien-6-one, trans,trans-3,7-Dimethyl-10-isopropylidene-3,7-cyclodecadien-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O
CAS Number:
Molecular Weight:
218.33
Beilstein/REAXYS Number:
2047453
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤5.0% water

format

neat

storage temp.

2-8°C

SMILES string

O=C1C(C/C=C(C)/CC/C=C(C)/C1)=C(C)C

InChI

1S/C15H22O/c1-11(2)14-9-8-12(3)6-5-7-13(4)10-15(14)16/h7-8H,5-6,9-10H2,1-4H3/b12-8+,13-7+

InChI key

CAULGCQHVOVVRN-SWZPTJTJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Germacrone, a sesquiterpene, is mainly present as a bioactive ingredient of various essential oils as well as Chinese traditional medicines.

Application

Germacrone may be used as an analytical standard for the determination of the analyte in Xingnaojing injection - a Chinese medicine, food products, and Smyrnium olusatyrum, Pimpinella anisum, Ledum groenlandicum essential oils by various chromatography techniques.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aromas from Quebec. IV. Chemical composition of the essential oil of Ledum groenlandicum: A review
Collin G
American Journal of Essential Oils and Natural Products, 2(3), 06-11 (2015)
Anti-tumor effect of germacrone on human hepatoma cell lines through inducing G2/M cell cycle arrest and promoting apoptosis
Liu Y, et al.
European Journal of Pharmacology, 698(1-3), 95-102 (2013)
Preparative isolation and purification of germacrone and curdione from the essential oil of the rhizomes of Curcuma wenyujin by high-speed counter-current chromatography
Yan J, et al.
Journal of Chromatography A, 1070(1-2), 207-210 (2005)
Germacrone inhibits the proliferation of breast cancer cell lines by inducing cell cycle arrest and promoting apoptosis
Zhong Z, et al.
European Journal of Pharmacology, 667(1-3), 50-55 (2011)
Combined assisted extraction techniques as green sample pre-treatments in food analysis
Moreda-Pi?eiro J and Moreda-Pi?eiro A
TrAC, Trends in Analytical Chemistry, 38(19), 3313-3323 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service