Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42046

Supelco

Robinetinidin chloride

analytical standard

Synonym(s):

3,3′,4′,5′,7-Pentahydroxyflavylium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClO6
CAS Number:
Molecular Weight:
322.70
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

assay

≥98.5% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

[Cl-].Oc1ccc2cc(O)c([o+]c2c1)-c3cc(O)c(O)c(O)c3

InChI

1S/C15H10O6.ClH/c16-9-2-1-7-3-12(19)15(21-13(7)6-9)8-4-10(17)14(20)11(18)5-8;/h1-6H,(H4-,16,17,18,19,20);1H

InChI key

SSFSVCKWRJIXDS-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service