Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41329

Supelco

1-Butanethiol

analytical standard

Synonym(s):

Butyl mercaptan, Mercaptan C4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3SH
CAS Number:
Molecular Weight:
90.19
Beilstein/REAXYS Number:
1730908
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.1 (vs air)

vapor pressure

83 mmHg ( 37.7 °C)

assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.443 (lit.)

bp

98 °C (lit.)

mp

−116 °C (lit.)

density

0.842 g/mL at 25 °C (lit.)

suitability

conforms to structure for H-NMR

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCS

InChI

1S/C4H10S/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

WQAQPCDUOCURKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

53.6 °F - closed cup

flash_point_c

12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service