40785
Caffeic acid 1,1-dimethylallyl ester
analytical standard
Synonym(s):
1,1-Dimethylallyl caffeate, 1,1-Dimethylallyl caffeic acid ester, 3-Methyl-2-butenyl caffeate
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥98.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
format
neat
SMILES string
C\C(C)=C\COC(=O)\C=C\c1ccc(O)c(O)c1
InChI
1S/C14H16O4/c1-10(2)7-8-18-14(17)6-4-11-3-5-12(15)13(16)9-11/h3-7,9,15-16H,8H2,1-2H3/b6-4+
InChI key
TTYOHMFLCXENHR-GQCTYLIASA-N
Application
Packaging
Recommended products
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service