40728
1-Iodo-2,2-dimethylpropane
technical, ≥95% (GC)
Synonym(s):
2,2-Dimethyl-1-iodopropane, Neopentyl iodide
About This Item
Recommended Products
grade
technical
assay
≥95% (GC)
contains
silver wool as stabilizer
refractive index
n20/D 1.489 (lit.)
n20/D 1.489
bp
70-71 °C/100 mmHg (lit.)
density
1.494 g/mL at 25 °C (lit.)
SMILES string
CC(C)(C)CI
InChI
1S/C5H11I/c1-5(2,3)4-6/h4H2,1-3H3
InChI key
CJTZXIJETZZARD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
91.4 °F - closed cup
flash_point_c
33 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service