Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

39718

Supelco

2,3-Dimethyl-1,3-butadiene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)C(CH3)=CH2
CAS Number:
Molecular Weight:
82.14
Beilstein/REAXYS Number:
605285
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

vapor pressure

269 mmHg ( 37.7 °C)

assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

contains

~0.01% 2,6-di-tert-butyl-4-methylphenol as stabilizer

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.438 (lit.)
n20/D 1.439

bp

68-69 °C (lit.)

mp

−76 °C (lit.)

density

0.726 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

neat

storage temp.

2-8°C

SMILES string

CC(=C)C(C)=C

InChI

1S/C6H10/c1-5(2)6(3)4/h1,3H2,2,4H3

InChI key

SDJHPPZKZZWAKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

30.2 °F - closed cup

flash_point_c

-1 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service